Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-phosphoinositide dependent protein kinase-1 (A02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PDPK1.

Manufacturer:  Abnova Corporation H00005170A02

Catalog No. 89-004-232


Add to cart

Description

Description

Sequence: ILKMGPVDKRKGLFARRRQLLLTEGPHLYYVDPVNKVLKGEIPWSQELRPEAKNFKTFFVHTPNRTYYLMDPSGNAHKWCRKIQEVWRQRYQSHPDAAVQ
Specifications

Specifications

3-phosphoinositide dependent protein kinase-1
Unconjugated
50% glycerol
NM_002613
MGC20087|MGC35290|PDK1|PRO0461
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant PDPK1.
PDPK1
Serum
PDPK1
PDPK1 (NP_002604, 457 a.a. ~ 556 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5170
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit