Learn More
PDX1 (goat) Goat anti-Mouse, Human, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Manufacturer:  MilliporeSigma 061385
Catalog No. 06-138-5MI
Add to cart
Description
Description
Anti-PDX1 (goat) Antibody is a Goat Polyclonal Antibody for detection of PDX1 also known as pancreatic and duodenal homeobox 1, somatostatin transcription factor 1, Insulin promoter factor 1 and has been validated in western blotting and immunohistochemistry.
Specifications
Specifications
PDX1 (goat)
Unconjugated
Serum
Goat
Unpurified
−20°C in undiluted aliquotes for up to 12 months
Polyclonal
Human, Murine
Immunohistochemistry, Western Blot
Unpurified goat polyclonal serum containing 0.05% sodium azide.
GSF; IDX-1; STF-1; IUF1; PDX-1; MODY4; IPF1; IUF-1; IPF-1
Recombinant protein corresponding to the N-terminus of mouse PDX1.
100μL
Primary
NP_032840