Learn More
PDX1 (rabbit) Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061379
Catalog No. 06-137-9MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-PDX1 (rabbit) Antibody is a Rabbit Polyclonal Antibody for detection of PDX1 also known as pancreatic and duodenal homeobox 1, somatostatin transcription factor 1, Insulin promoter factor 1 and has been validated in western blotting and immunohistochemistry.
Specifications
Specifications
PDX1 (rabbit)
Unconjugated
Serum
Rabbit
IgG
100μL
Primary
NP_032840
Immunohistochemistry, Western Blot
Unpurified rabbit polyclonal containing 0.05% sodium azide.
GSF; IDX-1; STF-1; IUF1; PDX-1; MODY4; IPF1; IUF-1; IPF-1
GST-tagged recombinant protein corresponding to the N-terminus of mouse PDX1.
Unpurified
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine