Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PDX1 (goat), Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 061385/DEL

Catalog No. 50-171-997

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Transcription factor PDX1, also known as pancreatic and duodenal homeobox 1, activates expression of the insulin and somatostatin genes. This protein is a key regulator of islet peptide hormone expression and also plays an essential role in pancreatic development. It has been shown to interact with the basic helix-loop-helix domains of TCF3(E47) and NEUROD1 and with HMG-I(Y). This protein also interacts with the methyltransferase SETD7. Mutations in this gene may be involved in several disorders of the pancreas or in diabetes mellitus.

Host Species: Goat
Cross Reactivity: Human, Mouse
Immunogen: Recombinant protein corresponding to the N-terminus of mouse PDX1.

Immunohistochemistry, Western Blotting

Specifications

Specifications

PDX1 (goat)
Serum
Recombinant protein corresponding to the N-terminus of mouse PDX1.
Primary
Human, Murine
Unconjugated
Goat
100μL
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit