Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEAR1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523838
Description
PEAR1 is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation.Specifications
PEAR1 | |
Polyclonal | |
PEAR1 | |
JEDI, MEGF12, hPEAR1, multiple EGF-like domains protein 12, multiple EGF-like-domains 12, multiple epidermal growth factor-like domains protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375033 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q5VY43 | |
PEAR1 | |
KLH conjugated synthetic peptide between 961-990 amino acids from the C-terminal region of human PEAR1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction