Learn More
PEBP1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3211
Catalog No. PA3211
Add to cart
Description
Description
PA3-211 detects PEBP1 in human, mouse, and rat samples. PA3-211 has successfully been used in Western blot, immunocytochemical, and immunohistochemical (paraffin) procedures. The PA3-211 immunogen is a synthetic peptide corresponding to the C-terminal amino acids C(167)YQAEWDDYVPKLYEQLSGK(187) of PEBP1
Specifications
Specifications
PEBP1
Polyclonal
whole serum with 0.02% sodium azide
P30086, P70296, P31044
Phosphatidylethanolamine-binding protein 1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PEBP1
Liquid
PEBP1
Synthetic peptide corresponding to the C-terminal amino acids C(167)YQAEWDDYVPKLYEQLSGK(187) of PEBP1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23980, 29542, 5037