Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pellino 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10056672SS
Description
Pellino 1 Polyclonal specifically detects Pellino 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pellino 1 | |
Polyclonal | |
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:100 | |
DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1 | |
Rabbit | |
Unpurified | |
RUO | |
57162 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide | |
PELI1 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 100-150 of human Pellino 1 (Peli1), Genbank No. NP_065702.2. | |
0.05 mL | |
Primary | |
Human, Mouse | |
Antisera |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction