Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pellino 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$204.00 - $482.50

Specifications

Antigen Pellino 1
Dilution Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:100
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056672T
SDP
View Documents
Novus Biologicals
NB10056672SS
0.05 mL
Each for $204.00
Add to Cart
 
NB10056672
SDP
View Documents
Novus Biologicals
NB10056672
0.2 mL
Each for $482.50
Add to Cart
 
Description

Description

Pellino 1 Polyclonal specifically detects Pellino 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Pellino 1
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1
PELI1
IgG
Unpurified
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:100
Polyclonal
Antisera
RUO
Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
57162
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 100-150 of human Pellino 1 (Peli1), Genbank No. NP_065702.2.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit