Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520301
Description
A suggested positive control is K562 cell lysate. PA5-20301 can be used with blocking peptide PEP-0426.
PEN2 Polyclonal antibody specifically detects PEN2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
PEN2 | |
Polyclonal | |
Unconjugated | |
PSENEN | |
1700023M09Rik; gamma-secretase subunit PEN-2; hematopoietic stem/progenitor cells protein MDS033; liver regeneration-related protein LRRGT00140; LRRGT00140 mRNA; MDS033; MSTP064; Pen2; Pen-2; presenilin enhancer 2; presenilin enhancer 2 homolog; presenilin enhancer 2 homolog (C. elegans); presenilin enhancer gamma secretase subunit; presenilin enhancer gamma-secretase subunit; presenilin enhancer protein 2; presenilin enhancer, gamma-secretase subunit; Psenen; RGD1312037 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
292788, 55851, 66340 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6QI68, Q9CQR7, Q9NZ42 | |
PSENEN | |
A 13 amino acid peptide from near the carboxy terminus of human PEN2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction