Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentraxin 3/TSG-14 Mouse anti-Human, Clone: 247911, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen Pentraxin 3/TSG-14
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pentraxin 3/TSG-14 Glu18-Ser381 Accession # P26022
Clone 247911
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1826SP
View Documents
R&D Systems
MAB1826SP
25 ug
Each for $119.60
Add to cart
 
MAB1826100
View Documents
R&D Systems
MAB1826100
100 ug
Each for $377.00
Add to cart
 
MAB1826500
View Documents
R&D Systems
MAB1826500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Pentraxin 3/TSG-14 Monoclonal antibody MAB1826 Specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for ELISA Capture
Specifications

Specifications

Pentraxin 3/TSG-14
247911
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
5806
RUO
Mouse myeloma cell line NS0-derived recombinant human Pentraxin 3/TSG-14 Glu18-Ser381 Accession # P26022
ELISA
Purified
P26022
Monoclonal
Primary
Detects human Pentraxin 3/TSG-14 in ELISAs.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit