Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PEPT2/SLC15A2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP147206

Catalog No. NBP147206


Add to cart

Description

Description

PEPT2/SLC15A2 Polyclonal antibody specifically detects PEPT2/SLC15A2 in Human, Porcine samples. It is validated for Peptide ELISA.
Specifications

Specifications

PEPT2/SLC15A2
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
SLC15A2
Peptide with sequence C-QGNMIKLETKKTK corresponding to C-Terminus according to NP_066568.2.
Immunogen affinity purified
RUO
Primary
6565
Human, Porcine
ELISA
Peptide ELISA Detection limit 1:8000
NP_066568.2
Kidney H(+)/peptide cotransporter, kidney isoform, PEPT2FLJ33407, Peptide transporter 2, solute carrier family 15 (H+/peptide transporter), member 2, solute carrier family 15 member 2
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific to the C Terminus.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only