Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PER1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1524

Catalog No. PA1524

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-524 detects PER1 from human and mouse tissues as well as recombit mouse PER1. PA1-524 has been sucessfully used in Western blot, immunofluoresence and immunohistochemistry procedures. By Western blot, this antibody detects an ~136 kDa protein representing overexpressed recombit mouse PER1 protein. Immunofluorescence staining of PER1 in mouse testes with PA1-524 results in cytoplasmic staining. PA1-524 immunizing peptide corresponds to amino acid residues 39-51 from mouse PER1. PA1-524 immunizing peptide (Cat. No. PEP-076) is available for use in neutralization and control experiments.

Circadian rhythmicity is a basic property of phylogenetically diverse organisms which range from animals and plants, to fungi. Regulation of endogenous biological clocks is regulated at the genetic level by a protein-mediated, autoregulatory feed-back loop. In mammals, several genes that encode members of the basic helix-loop helix (bHLH) PAS (PER-ARNT-SIM) transcription factor family have been shown to play a significant role in regulating circadian oscillations. Transactivation of CLOCK-induced genes is mediated via an E box enhancer (CACGTG) found upstream of target genes. CLOCK-ARNT3 heterodimers bind to E box regulatory elements and stimulate gene transcription. CLOCK has been shown to transactivate the mammalian homolog of Drosophila per. PER, in concert with the product of the mammalian timeless gene (TIM), negatively regulates its own transcription by blocking the activity of the CLOCK-BMAL1 transactivation complex.
Specifications

Specifications

PER1
Polyclonal
Unconjugated
PER1
Liquid
PER1
Synthetic Peptide: P(39) S L A D D T D A N S N G(51) C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O15534, O35973
Period 1
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18626, 5187
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit