Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Per2 Mouse, Unlabeled, Clone: 52, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611138

Catalog No. BDB611138


Add to cart

Description

Description

Circadian rhythms are behavioral or physiological rhythms with periods of approximately 24 hours. They have been identified in many species from bacteria to humans. In mammals, the circadian regulator is the suprachiasmatic nucleus (SCN) of the anterior hypothalamus. Multiple genes involved in the SCN circadian rhythm have been isolated. The Drosophila period (dper) gene is the most completely characterized mammalian clock gene. In mouse, multiple per genes have been identified and constitute a mammalian period gene family (mper1, mper2, and mper3). These proteins contain a dimerization PAS domain and a cytoplasmic localization domain. SCN expression of mper1 and mper2 overlaps, but is not synchronous. Although not light-inducible, mper2 transcripts are detected approximately two hours following those of mper1. This suggests that the mper1 gene product, a putative transcription factor, is produced in response to light and promotes the expression of mper2. Although mper1 and mper2 are similar, they are not redundant. Thus, these different circadian regulators may allow integration of a wider range of input signals and interact with different downstream effectors.

Host Species: Mouse
Clone: 52
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse Per2 aa. 1-112
Formula Weight [Chemical]: 105kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

Per2
52
Unconjugated
Affinity Purified
Mouse Per2 aa. 1-112
50μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit