Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521436

Catalog No. PIPA521436


Add to cart

Description

Description

PA5-21436 targets Perforin 1 in WB applications and shows reactivity with Human samples. The PA5-21436 immunogen is a recombit fragment corresponding to a region within amino acids 9 and 214 of Perforin 1 (Uniprot IDNo.P14222). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
Specifications

Specifications

Perforin
Polyclonal
Unconjugated
PRF1
Liquid
PRF1
Recombit fragment corresponding to a region within amino acids 9 and 214 of Perforin 1 (Uniprot IDNo.P14222)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.37 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P14222
RP11-710A11.3, FLH2, HPLH2, P1, PFN1, PFP, cytolysin, lymphocyte pore forming protein, lymphocyte pore-forming protein, perforin-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5551
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit