Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP85 Goat anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131346

Catalog No. PIPA131346


Add to cart

Description

Description

PA1-31346 detects Pericentrin 1 from human, mouse, and rat samples. PA1-31346 has been successfully used in Western blot and ELISA applications. Positive control of HeLa lysate suggested. The PA1-31346 immunogen is synthetic peptide: ARAIIREGSLEGS, corresponding to C terminal amino acids 644-656 of Human Pericentrin1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

NUP85
Polyclonal
Unconjugated
NUP85
Liquid
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
287830, 445007, 79902
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9BW27, Q8R480, Q4QQS8
NUP85
synthetic peptide ARAIIREGSLEGS, corresponding to C terminal amino acids 644-656 of Human Pericentrin1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit