Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peripherin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA110018
Description
PA1-10018 was made against full length human peripherin.| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Peripherin Polyclonal antibody specifically detects Peripherin in Bovine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Peripherin | |
Polyclonal | |
Unconjugated | |
PRPH | |
if3; nef4; Neurofilament 4; neurofilament 4 (57kD); neuronal intermediate filament IF3; Perf; Peripherin; peripherin 1; peripherin L homeolog; plasticin; PRPH; prph.L; PRPH1; XELAEV_18013332mg; XIF3 | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 5mM sodium azide | |
A6QQJ3, P15331, P21807, P41219 | |
PRPH | |
Purified, recombinant human peripherin, expressed in E. coli. | |
RUO | |
100152434, 19132, 24688, 510082, 5630 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction