Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peripherin Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA110012
Description
Stable at 4°C for at least one year. Mix 1:1 with 100% glycerol and store at -20°C for longer term storage.
Peripherin Polyclonal antibody specifically detects Peripherin in Bovine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Peripherin | |
Polyclonal | |
Unconjugated | |
PRPH | |
if3; nef4; Neurofilament 4; neurofilament 4 (57kD); neuronal intermediate filament IF3; Perf; Peripherin; peripherin 1; peripherin L homeolog; plasticin; PRPH; prph.L; PRPH1; XELAEV_18013332mg; XIF3 | |
Chicken | |
PEG precipitation | |
RUO | |
100152434, 19132, 24688, 510082, 5630 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Conc. Not Determined | |
PBS with 0.02% sodium azide | |
A6QQJ3, P15331, P21807, P41219 | |
PRPH | |
Recombinant rat peripherin expressed in and purified from E. coli | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction