Learn More
PRDX3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3752
Catalog No. PA3752
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
PA3-752 detects Peroxiredoxin 3 protein in human samples. This antibody is specific for Prx 3 and shows no cross-reactivity with other Prx isoforms. PA3-752 has successfully been used in Western blot, and immunocytochemistry procedures. By Western blot, this antibody detects an ~27 kDa protein representing Prx 3 in human PC3 cell lysate. The PA3-752 antigen is purified human Recombinant Prx 3 from HEK 293 cells.
The peroxiredoxin (Prdx, Prx, or Trx-Px) family of enzymes is a recently identified family of peroxidases found in free-living organisms. The six known isoforms (Prx1-6) play an important role in protecting lipids, enzymes and DNA against peroxides, such as hydrogen peroxide. The ubiquitously expressed peroxiredoxins have also been shown to play a role in apoptosis and cell differentiation. This is acomplished by the active cysteine of Prx reducing peroxides, which is then converted into a transient cysteine sulfenic acid or cysteine sulfinic acid. If Prx protection is inadequate against peroxidases, the resulting protein and DNA damage may result in neurological disease such as Alzheimer’s or DNA damage leading to cancer.
Specifications
Specifications
PRDX3
Polyclonal
whole serum with 0.05% sodium azide
P30048
Prx 3
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
PRDX3
Liquid
PRDX3
Purified human recombit Peroxiredoxin 3.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10935