Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14284

Catalog No. PA14284


Add to cart

Description

Description

PA1-4284 detects PERP from human samples. PA1-4284 has been successfully used in Western blot procedures. PA1-4284 immunizing peptide corresponds to amino acid residues 37-50 from human PERP protein.

The p53 tumor suppressor activates either cell cycle arrest or apoptosis in response to cellular stress. A novel gene, PERP, is expressed in a p53-dependent manner and at high levels in apoptotic cells compared with G(1)-arrested cells. PERP induction is linked to p53-dependent apoptosis and is directly activated by p53. These data suggest that PERP is a novel effector of p53-dependent apoptosis.
Specifications

Specifications

PERP
Polyclonal
Unconjugated
PERP
Liquid
PERP
Synthetic Peptide: Q(37) S S D H G Q T S S L W W K(50)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96FX8
p53 apoptosis effector related to PMP-22, Keratinocytes-associated protein 1, KCP-1, P53-induced protein PIGPC1, Transmembrane protein THW
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64065
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit