Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146283

Catalog No. PIPA146283


Add to cart

Description

Description

The p53 tumor suppressor activates either cell cycle arrest or apoptosis in response to cellular stress. A novel gene, PERP, is expressed in a p53-dependent manner and at high levels in apoptotic cells compared with G(1)-arrested cells. PERP induction is
Specifications

Specifications

PERP
Polyclonal
Unconjugated
PERP
Liquid
PERP
Amino acids 37-50 of the human PERP protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96FX8, Q9JK95
KCP1, KRTCAP1, PIGPC1, RP3-496H19.1, THW, dJ496H19.1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64058, 64065
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit