Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PES1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126573

Catalog No. PIPA126573

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26573 detects Pescadillo from mouse samples.PA1-26573 has been successfully used in Western blot procedures.The PA1-26573 immunogen is a synthetic peptide corresponding to a portion of mouse Pescadillo encoded in part by exons13 and 14.
Specifications

Specifications

Pescadillo
Polyclonal
Unconjugated
Pes1
Purified
Rabbit
IgG
50μL
Store at 4°C
Primary
64934
Immunocytochemistry, Immunofluorescence, Western Blot
1mg/mL
tris phosphate, tris citrate with 0.1% sodium azide; pH 7-8
Q9EQ61
PES1
Synthetic peptide corresponding to a portion of mouse Pescadillo encoded in part by exons13 and 14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit