Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX11A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526025

Catalog No. PIPA526025

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.
Specifications

Specifications

PEX11A
Polyclonal
PBS with 0.09% sodium azide
O75192
PEX11-ALPHA, 28 kDa peroxisomal integral membrane protein, PMP28, hsPEX11p, peroxin-11A, peroxisomal biogenesis factor 11A, peroxisomal membrane protein 11A, protein PEX11 homolog alpha
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8800
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PEX11A
Liquid
PEX11A
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PEX11A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit