Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PFKFB3.
Supplier: Abnova Corporation H00005209A01
Description
Sequence: CPLHTVLKLTPVAYGCRVESIYLNVESVCTHRERSEDAKKGPNPLMRRNSVTPLASPEPTKKPRINSFEEHVASTSAALPSCLPPEVPTQLPGQNMKGSRSSADSSRKHSpecifications
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PFKFB3. | |
PFKFB3 | |
IPFK2/PFK2 | |
Mouse | |
50 μL | |
Yes | |
5209 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004566 | |
PFKFB3 | |
PFKFB3 (NP_004557, 412 a.a. ∼ 520 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction