Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKFB3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521931

Catalog No. PIPA521931


Add to cart

Description

Description

PA5-21931 targets PFKFB3 in WB applications and shows reactivity with Human samples. The PA5-21931 immunogen is synthetic peptide contain a sequence corresponding to a region within amino acids 457 and 520 (Q16875) of PFKFB3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

PFKFB3
Polyclonal
Unconjugated
PFKFB3
Liquid
PFKFB3
Synthetic peptide contain a sequence corresponding to a region within amino acids 457 and 520 (Q16875) of PFKFB3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.95 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q16875
RP11-298K24.3, IPFK2, PFK2, 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase, 6-phosphofructo-2-kinase/fructose-2, 6-bisphosphatase, 6PF-2-K/Fru-2,6-P2ase 3, 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme, PFK/FBPase 3, fructose-6-phosphate,2-kinase/fructose-2, 6-bisphosphatase, iPFK-2, inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, renal carcinoma antigen NY-REN-56
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5209
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit