Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKFB4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528392

Catalog No. PIPA528392


Add to cart

Description

Description

PA5-28392 targets PFKFB4 in WB applications and shows reactivity with Human samples. The PA5-28392 immunogen is recombit fragment corresponding to a region within amino acids 160 and 454 of Human PFKFB4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Synthesis and degradation of fructose 2,6-bisphosphate.
Specifications

Specifications

PFKFB4
Polyclonal
Unconjugated
PFKFB4
Liquid
PFKFB4
Recombit fragment corresponding to a region within amino acids 160 and 454 of Human PFKFB4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q16877, Q6DTY7
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4, 6PF-2-K/Fru-2,6-P2ase 4, 6PF-2-K/Fru-2,6-P2ase testis-type isozyme, PFK/FBPase 4, bifunctional enzyme with kinase and biphosphatase activities
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
270198, 5210
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit