Learn More
anti-PGAM1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131074
Catalog No. NBP131074
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PGAM1
0.97mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PGAM1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PGAM1. The exact sequence is proprietary.
29 kDa
0.1 ml
Stem Cell Markers
Primary
5223
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%), Xenopus tropicalis (100%), Rhesus Monkey (100%), and Chimpanzee (100%). Not yet tested in any other species.