Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524006

Catalog No. PIPA524006

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and non-human primate based on sequence homology.

Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). PGAM2 encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase eficiency, also known as glycogen storage disease X.
Specifications

Specifications

PGAM2
Polyclonal
PBS with 0.09% sodium azide
P15259, O70250
GSD10, PGAM-M, PGAMM, BPG-dependent PGAM 2, muscle-specific phosphoglycerate mutase, phosphoglycerate mutase 2, phosphoglycerate mutase isozyme M
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5224, 56012
Immunohistochemistry, Western Blot
Unconjugated
PGAM2
Liquid
PGAM2
KLH conjugated synthetic peptide between 12-41 amino acids from the N-terminal region of human PGAM2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit