Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525635

Catalog No. PIPA525635

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with hamster, mouse and rat based on sequence homology.

PGAP2 is involved in the lipid remodeling steps of GPI-anchor maturation. Required for stable expression of GPI-anchored proteins at the cell surface.
Specifications

Specifications

PGAP2
Polyclonal
Unconjugated
PGAP2
Liquid
PGAP2
KLH conjugated synthetic peptide between 214-242 amino acids from the C-terminal region of human PGAP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9UHJ9
CWH43-N, FRAG1, FGF receptor activating protein 1, FGF receptor-activating protein 1, cell wall biogenesis 43 N-terminal homolog, post-GPI attachment to proteins factor 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27315
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit