Learn More
MilliporeSigma™ anti-PGC-1, Polyclonal, ™ (Chemicon™)
Supplier: MilliporeSigma™ AB3242/DEL
Description
PGC-1 is a transcription factor that determines whether preadipocytes will differentiate into brown or white fat cells. It interacts with PPAγ to promote the expression of a cassette of genes which define the brown adipocyte phenotype. GenBank Number: NM_013261. UniProt Number: Q9UBK2 (human). MW: 100kDa.
ELISA, Immunocytochemistry, Western Blotting
Specifications
PGC-1 | |
Unconjugated | |
Synthetic peptide from amino acids 777-797 (C-terminus) of mouse PGC-1 | |
100 μg | |
Epigenetics | |
Human, Mouse |
Polyclonal | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary |