Learn More
MilliporeSigma™ anti-PGC-1alpha Clone: 4C1.3, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST12021SET
Catalog No. ST12021SET
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifically detects PGC-1α Clone: 4C1.3 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
PGC-1α is a co-activator of PPARα, PPARγ, and other transcription factors and regulates the transcriptonal program of adaptive thermogenesis in brown adipose tissue, hepatic/renal gluconeogenesis, and muscle fiber type switching. The full-length protein is 113kDa and is induced in brown adipose tissue by cold exposure, in liver and kidney by fasting, and in skeletal muscle by exercise. Alternative splicing of the full-length gene produces a 270 aa N-terminal splice variant that migrates at ~38kDa and is induced by the same physiological signals that induce expression of the full-length protein. The N-terminal PGC-1α splice variant is also subject to post translational modifications that alter its migration and apparent molecular weight.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PGC-1α
4C1.3
One vial of 100μg antibody in 50mM PBS, see vial for lot-specific concentration. One vial of PGC-1α Positive Control and one vial of NT-PGC-1α Positive Control, supposed as 25μL whole cell extracts in RIPA buffer containing 10mM β-mercaptoethanol and 1% SDS; load 10μL per lane; add sample buffer prior to SDS-PAGE loading.
Murine
IgG
RUO
Primary
Recognizes the endogenous forms of PGC-1α, which includes the ∽113kDa PGC-1α protein and the ∽38kDa splice variant, in brown adipose tissue, liver, and kidney.
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Purified
a recombinant protein consisting of amino acids 1-120 of mouse PGC-1α
1 Set
≤ -70°C
Monoclonal
Human, Murine, Rat