Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGF Mouse anti-Human, Clone: 37203, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523734

Catalog No. PIMA523734


Add to cart

Description

Description

In ELISAs, this antibody does not cross-react with recombit human (rh) VEGF, rhPDGF-AA, rhPDGF-AB, or rhPDGF-BB. Reconstitute at 0.5 mg/mL in sterile PBS.

Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
Specifications

Specifications

PGF
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P49763
PGFL, PGF, Placental Growth Factor, PLGF, SHGC, Vascular Endothelial Growth Factor-Related Protein
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5228
ELISA
37203
Unconjugated
PGF
Lyophilized
PGF
E. coli-derived recombit human PlGF Ala21-Arg149
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit