Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGP9.5 (soluble) Guinea Pig anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Guinea Pig Polyclonal Antibody

Manufacturer:  Invitrogen PA124674

Catalog No. PA124674


Add to cart

Description

Description

PA1-24674 detects PGP9.5 from human, mouse, and rat samples. PA1-24674 is expected to cross react with Macaque monkey (87% conserved) due to sequence homology. PA1-24674 has been successfully used in immunohistochemistry (frozen) procedures. The PA1-24674 immunogen is a synthetic peptide corresponding to residues G(175) A S S E D T L L K D A A K V C R(191) of soluble, cytoplasmic, human PGP9.5. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze/thaw cycles.

PGP9.5 (Protein gene product 9.5, UCH-L1, PARK5) is a neuron specific protein, structurally and immunologically distinct from neuron specific enolase. PGP9.5 has a molecular weight of 27 kDa and was first identified by high resolution two dimensional PAGE. PGP9.5 is a member of ubiquitin carboxyl-terminal hydrolase family 1 (peptidase family C12) with a ubiquitin carboxyl-terminal hydrolase domain. PGP9.5 is well known for having ubiquitin hydrolase and ligase activities that hydrolyzes small C-terminal adducts of ubiquitin to generate ubiquitin monomers. PGP9.5 is present in neurons and nerve fibers at all levels of the central and peripheral nervous system, in neuroendocrine cells, in segments of the renal tubules, in spermatogonia and Leydig cells of the testis, in ova and in some cells of both the pregt and non-pregt corpus luteum. Over expression of PGP9.5 leads to non-small cell lung cancer while decreased expression leads to Huntington disease and Alzheimer disease. Since PGP9.5 is present in cellular inclusions, it can be a useful as a neuronal marker and in the studies of neurodegenerative disorders such as with Parkinson disease.
Specifications

Specifications

PGP9.5 (soluble)
Polyclonal
whole serum with 0.05% sodium azide
P09936, Q9R0P9, Q00981
PARK5, PGP9.5, PGP95, Uch-L1
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
UCHL1
Liquid
UCHL1
Synthetic peptide corresponding to residues G(175) A S S E D T L L K D A A K V C R(191) of soluble, cytoplasmic, human PGP9.5.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22223, 29545, 7345
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit