Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGPEP1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528364

Catalog No. PIPA528364


Add to cart

Description

Description

PA5-28364 targets PGPEP1 in WB applications and shows reactivity with mouse samples. The PA5-28364 immunogen is recombit fragment corresponding to a region within amino acids 1 and 209 of Human PGPEP1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Pyroglutamyl peptidase I (EC 3.4.19.3) catalyzes the hydrolysis of N-terminal pyroglutamyl residues from oligopeptides and proteins.
Specifications

Specifications

PGPEP1
Polyclonal
Unconjugated
Pgpep1
Liquid
PGPEP1
Recombit fragment corresponding to a region within amino acids 1 and 209 of Human PGPEP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q9ESW8
PGP, PGP-I, PGPI, Pcp, 5-oxoprolyl-peptidase, PAP-I, pyroglutamyl aminopeptidase I, pyroglutamyl-peptidase 1, pyrrolidone-carboxylate peptidase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
66522
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit