Learn More
Phafin 1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA124323
Catalog No. PIPA124323
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
PA1-24323 detects APPD in human samples.PA1-24323 has been successfully used in Western blot procedures.PA1-24323 immunogen is a synthetic peptide to a region of human PLEKHF1.
Specifications
Specifications
Phafin 1
Polyclonal
Unconjugated
PLEKHF1
Purified
PLEKHF1
Synthetic peptide to a region of human PLEKHF1.
Protein A/G
RUO
Antibody
Polyclonal
Human
Western Blot
1mg/mL
PBS with 2% sucrose and no preservative
Q96S99
AI037048, SGP-1
Rabbit
IgG
100μg
Avoid Freeze/Thaw Cycles
Primary
79156