Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHAP I Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130728

Catalog No. PIPA130728


Add to cart

Description

Description

PA1-30728 detects PHAP1 from mouse, human, rat samples. PA1-30728 has been successfully used in Western blot applications. The PA1-30728 immunogen is: Synthetic peptide (Human), corresponding to amino acids at carboxy terminus of human PHAP I. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

PHAP I
Polyclonal
Unconjugated
ANP32A
Liquid
ANP32A
Synthetic peptide (Human), corresponding to amino acids at carboxy terminus of human PHAP I.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P39687, O35381, P49911
C15orf1, I1PP2A, LANP, MAPM, MGC119787, MGC150373, PHAP1, PHAPI, PP32
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11737, 25379, 8125
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit