Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHAPI2, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131866

Catalog No. PIPA131866

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

APRIL (a proliferation-inducing ligand) is a newly identified member of the tumor necrosis factor (TNF) family. Ectopic expression of APRIL is associated with the upregulation of X-linked inhibitor of apoptosis protein (XIAP) and may act as antiapoptotic

PA1-31866 detects PHAPI2 from human samples.PA1-31866 has been successfully used in Western blot applications.The PA1-31866 immunogen is: Peptide with sequence KRKRETDDEGEDD, from C Terminus of the protein sequence according to NP_006392
Specifications

Specifications

PHAPI2
Polyclonal
Unconjugated
ANP32B
Purified
ANP32B
Peptide with sequence KRKRETDDEGEDD, from C Terminus of the protein sequence according to NP_006392
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide
Q92688, Q9EST5, Q9EST6
APRIL
Goat
IgG
50μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10541
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit