Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512275

Catalog No. PIPA512275


Add to cart

Description

Description

This antibody is predicted to react with bovine and chicken based on sequence homology.

Prohibitin is an evolutionarily conserved protein that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3 ft. untranslated region.
Specifications

Specifications

Prohibitin
Polyclonal
PBS with 0.09% sodium azide
P35232
Prohibitin; PHB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5245
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
PHB
Liquid
PHB
KLH conjugated synthetic peptide between 89-117 amino acids from the central region of human PHB
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit