Learn More
PHD2 Mouse anti-Human, Clone: 76a, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051327
Catalog No. 05-132-7MI
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-PHD2 Antibody, clone 76a (mouse monoclonal antibody) validated in western blotting to detect PHD2 also known as EGL nine (C.elegans) homolog 1, HIF prolyl hydroxylase 2, Hypoxia-inducible factor prolyl hydroxylase 2.
Specifications
Specifications
PHD2
Monoclonal
Unconjugated
NP_071334.1
Mouse
Protein G purfied
Primary
2-8°C for one year
IgG1 κ
Western Blot
76a
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
EGLN1, C1orf12, ECYT3, HIF-PH2, HIFPH2, HPH-2, HPH2, PHD2, SM-20, SM20, ZMYND6
Recombinant protein corresponding to human PHD2.
100 μg
Human
Purified