Learn More
PHD3 Mouse anti-Human, Clone: 188e, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 051328
Catalog No. 05-132-8MI
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Anti-PHD3 Antibody, clone 188e is a mouse monoclonal antibody for detection of PHD3 also known as EGL nine (C.elegans) homolog 3, HIF-prolyl hydroxylase 3, Hypoxia-inducible factor prolyl hydroxylase 3, and has been validated in western blotting.
Specifications
Specifications
PHD3
Monoclonal
Unconjugated
NP_071356.1
Mouse
Protein G purfied
Primary
2-8°C for one year
IgG1 κ
Western Blot
188e
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
EGLN3, HIF-PH3, HIFPH3, HPH-1, PHD3
Recombinant protein corresponding to human PHD3.
100 μg
Human
Purified
Product Suggestions
Product Suggestions