Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PHD4/HIF Prolyl Hydroxylase 4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 46020002

Catalog No. 46-020-002


Add to cart

Description

Description

PHD4/HIF Prolyl Hydroxylase 4 Polyclonal antibody specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PHD4/HIF Prolyl Hydroxylase 4
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
P4HTM
This antibody is specific for the N Terminus Region of the target protein.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human PHD4/HIF Prolyl Hydroxylase 4
ELISA, Western Blot
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
Q9NXG6
EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase
Rabbit
IgG
0.05 mg
Cancer, Hypoxia, Lipid and Metabolism
Primary
54681
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only