Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN32, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131554

Catalog No. PIPA131554

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in malignancies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.

PA1-31554 detects PHEMX from human samples.PA1-31554 has been successfully used in Western blot applications.The PA1-31554 immunogen is: Peptide with sequence GGLSGCPERGLSD, from C Terminus of the protein sequence according to NP_005696; NP_620591
Specifications

Specifications

PHEMX
Polyclonal
Unconjugated
TSPAN32
Purified
TSPAN32
Peptide with sequence GGLSGCPERGLSD, from C Terminus of the protein sequence according to NP_005696; NP_620591
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q96QS1
ART1, FLJ17158, FLJ97586, MGC22455, PHEMX, PHMX, TSSC6
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
10077
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit