Learn More
Phenytoin Mouse anti-Chemical, Clone: PY25-512.4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110740
Catalog No. 01-671-970
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10740 detects Phenytoin. MA1-10740 has been successfully used in ELISA applications. The MA1-10740 immunogen is Phenytoin, conjugated to KLH.
Phenytoin sodium is a commonly used antiepileptic. Phenytoin (fen-i-TOE-in) acts to suppress the abnormal brain activity seen in seizure by reducing electrical conductance among brain cells by stabilizing the inactive state of voltage-gated sodium channels. Aside from seizures, it is an option in the treatment of trigeminal neuralgia in the event that carbamazepine is deemed inappropriate.
Specifications
Specifications
Phenytoin
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Phenytek; Dilantin; Kapseals; Infatabs; Epanutin
Phenytoin, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
PY25-512.4
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical