Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD finger protein 16, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PHF16 protein.

Manufacturer:  Abnova Corporation H00009767B01

Catalog No. 89-006-839


Add to cart

Description

Description

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq

Sequence: MKRHRPVSSSDSSDESPSTSFTSGSMYRIKSKIPNEHKKPAEVFRKDLISAMKLPDSHHINPDSYYLFADTWKEEWEKGVQVPASPDTVPQPSLRIIAEKVKDVLFIRPRKYIHCSSPDTTEPGYINIMELAASVCRYDLDDMDIFWLQELNEDLAEMGCGPVDENLMEKTVEVLERHCHENMNHAIETEEGLGIEYDEDVICDVCRSPDSEEGNDMVFCDKCNVCVHQACYGILKVPEGSWLCRSCVLGIYPQCVLCPKKGGALKTTKTGTKWAHVSCALWIPEVSIACPERMEPITKISHIPPSRWALVCNLCKLKTGACIQCSIKSCITAFHVTCAFEHGLEMKTILDEGDEVKFKSYCLKHSQNRQKLGEAEYPHHRAKEQSQAKSEKTSLRAQKLRELEEEFYSLVRVEDVAAELGMPTLAVDFIYNYWKLKRKSNFNKPLFPPKEDEENGLVQPKEESIHTRMRMFMHLRQDLERVRNLCYMISRREKLKLSHNKIQEQIFGLQVQLLNQEIDAGLPLTNALENSLFYPPPRITLKLKMPKSTPEDHRNSSTETDQQPHSPDSSSSVHSIRNMQVPQESLEMRTKSYPRYPLESKNNRLLASLSHSRSEAKESSPAWRTPSSECYHGQSLGKPLVLQAALHGQSSIGNGKSQPNSKFAKSNGLEGSWSGNVTQKDSSSEMFCDQEPVFSPHLVSQGSFRKSTVEHFSRSFKETTNRWVKNTEDLQCYVKPTKNMSPKEQFWGRQVLRRSAGRAPYQENDGYCPDLELSDSEAESDGNKEKVRVRKDSSDRENPPHDSRRDCHGKSKTHPLSHSSMQR
Specifications

Specifications

PHD finger protein 16
Unconjugated
No additive
NM_014735.2
JADE3|KIAA0215|MGC138748|MGC138749
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9767
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human PHF16 protein.
PHF16
Serum
PHF16
PHF16 (NP_055550.1, 1 a.a. ~ 823 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit