Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528504

Catalog No. PIPA528504


Add to cart

Description

Description

PA5-28504 targets PHF17 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28504 immunogen is recombit protein fragment corresponding to a region within amino acids 298 and 536 of Human PHF17. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

PHF17
Polyclonal
Unconjugated
JADE1
Liquid
JADE1
Recombit protein fragment corresponding to a region within amino acids 298 and 536 of Human PHF17
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.77 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q6IE81
JADE1, PHD protein Jade-1, gene for apoptosis and differentiation in epithelia, protein Jade-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
79960
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit