Learn More
anti-PHF20, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132464
Catalog No. NBP132464
Add to cart
Description
Description
PHF20 Polyclonal antibody specifically detects PHF20 in Human samples. It is validated for Western Blot.
Specifications
Specifications
PHF20
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PHF20
Recombinant protein encompassing a sequence within the N-terminus region of human PHD finger protein 20. The exact sequence is proprietary.
115 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
GLEA2, PHD finger protein 20
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51230
Human