Learn More
PHGDH Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524633
Catalog No. PIPA524633
Add to cart
Description
Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
Specifications
Specifications
PHGDH
Polyclonal
Unconjugated
PHGDH
Liquid
PHGDH
KLH conjugated synthetic peptide between 249-277 amino acids from the central region of human PHGDH
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
O43175
3-PGDH, 3PGDH, PDG, PGAD, PGD, PGDH, SERA, 3-phosphoglycerate dehydrogenase, D-3-phosphoglycerate dehydrogenase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26227