Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHLPP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514908

Catalog No. PIPA514908

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PHLPP2 is a protein phosphatase that specifically mediates dephosphorylation of  ft.Ser-473 ft. of AKT1. This protein regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of  ft.Ser-473 ft. of AKT1 triggers apoptosis and decrease cell proliferation. It also controls the phosphorylation of AKT3.
Specifications

Specifications

PHLPP2
Polyclonal
PBS with 0.09% sodium azide
Q6ZVD8
PH domain leucine-rich repeat protein phosphatase-like; PHLPP-like; PHLPP2; PHLPPL; KIAA0931
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23035
Western Blot
Unconjugated
PHLPP2
Liquid
PHLPP2
KLH conjugated synthetic peptide between 4-31 amino acids from the N-terminal region of human PHLPP2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit