Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Hydroxylase (Ser31) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Catalog No. 01674870
Change view
Click to view available options
Quantity:
10 Miniblots
Catalog No. Quantity
01-674-870 10 Miniblots
1 options

Catalog No. 01-674-870

Supplier: Thermo Scientific PA112745

Rabbit Polyclonal Antibody

Tyrosine Hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines Dopamine and Norepinephrine. TH antibodies can therefore be used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson’s disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999). TH antibodies can also be used to explore basic mechanisms of dopamine and norepinephrine signaling (Witkovsky et al., 2000; Salvatore et al., 2001; Dunkley et al., 2004). The activity of TH is also regulated by phosphorylation (Haycock et al., 1982; Haycock et al., 1992; Jedynak et al., 2002). Phospho-specific antibodies for the phosphorylation sites on TH can be used to great effect in studying this regulation and in identifying the cells in which TH phosphorylation occurs.

A suggested positive control for this product is PC-12 cell lysate stimulated with okadaic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Tyrosine Hydroxylase (Ser31)
Applications Dot Blot, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene TH
Gene Accession No. P07101, P24529, P04177
Gene Alias TH, TYH antibody, Tyrosine 3 monooxygenase antibody
Gene Symbols TH
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to amino acid residues surrounding phospho-Ser31 of rat tyrosine hydroxylase
Purification Method Affinity chromatography
Quantity 10 Miniblots
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21823, 25085, 7054
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.