Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-14-3-3 beta (Ser58) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14612
Description
This antibody is predicted to react with bovine, canine, chicken, mouse, non-human primate, sheep, Xenopus and zebrafish based on 100% sequence homology. This antibody is specific for the ∽29 kDa 14-3-3 protein phosphorylated at Ser58. Immunolabeling is blocked by preadsorption of the antibody with the protein used to generate the antibody.
Phospho-14-3-3 beta (Ser58) Polyclonal antibody specifically detects Phospho-14-3-3 beta (Ser58) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Phospho-14-3-3 beta (Ser58) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P31946, P35213, Q9CQV8 | |
YWHAB | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser58 of rat YWHAB conjugated to KLH | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
YWHAB | |
1300003C17Rik; 14-3-3 alpha; 14-3-3 protein beta; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha, N-terminally processed; 14-3-3 protein beta-subtype; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; brain protein 14-3-3, beta isoform; epididymis secretory protein Li 1; GW128; HEL-S-1; HS1; hypothetical protein GW128; KCIP 1; KCIP1; KCIP-1; Prepronerve growth factor RNH-1; protein 1054; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; QccE-16215; RP1-148E22.1; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3/tryptophan 5 -monooxygenase activation protein beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation pro | |
Rabbit | |
Affinity chromatography | |
RUO | |
54401, 56011, 7529 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction