Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-AKT1 (Ser473) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137046

Catalog No. 01-673-030


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.

The serine threonine protein kinase encoded by the AKT-1 gene is catalytically inactive in serum starved primary and immortalized fibroblasts. AKT-1 and the related AKT-2 are activated by platelet derived growth factor. The activation is rapid and specific. In the developing nervous system AKT is a critical mediator of growth factor induced neuronal survival. Survival factors can suppress apoptosis in a transcription independent manner by activating the serine/threonine kinase AKT-1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene (referenced from entrez gene). The antibody labels activated AKT-1 that is phosphorylated at 473 serine residue.
Specifications

Specifications

Phospho-AKT1 (Ser473)
Polyclonal
PBS with 1% BSA and 0.099% sodium azide; pH 7.6
P31749
AKT, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA, PKB alpha, RAC-PK-alpha, RAC-alpha serine/threonine-protein kinase, protein kinase B alpha, proto-oncogene c-Akt, rac protein kinase alpha
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
207
Immunohistochemistry (Paraffin)
Unconjugated
AKT1
Liquid
AKT1
Synthetic peptide derived from the region of human AKT-1 that contains phosphorylated site of serine 473.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit